siRNA Details
| Gene Symbol: NAPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013975.1 | XP_016869464.1 | 11 | 3448 | ||
| XM_017013977.1 | XP_016869466.1 | 11 | 3148 | |||
| XM_017013978.1 | XP_016869467.1 | 10 | 3100 | |||
| XM_017013979.1 | XP_016869468.1 | 11 | 1644 | |||

NAPRT
Genomic Map