siRNA Details
Gene Symbol: LOC100131174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130404.1 | - | 1 | 1281 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100131174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130404.1 | - | 1 | 1281 |