siRNA Details
| Gene Symbol: PQLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026002.1 | XP_016881491.1 | 4 | 515 | ||
| GenBank mRNA | AK126188.1 | - | 1 | 537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PQLC1| Gene Symbol: PQLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026002.1 | XP_016881491.1 | 4 | 515 | ||
| GenBank mRNA | AK126188.1 | - | 1 | 537 | ||