siRNA Details
| Gene Symbol: LOC100996573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003846334.4 | XP_003846382.2 | 1 | 1698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996573| Gene Symbol: LOC100996573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003846334.4 | XP_003846382.2 | 1 | 1698 | ||