siRNA Details
| Gene Symbol: LOC101926934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134643.1 | 1, 2 | 233 | |||
| GenBank mRNA | AI698692.1 | - | 1, 2 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926934| Gene Symbol: LOC101926934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134643.1 | 1, 2 | 233 | |||
| GenBank mRNA | AI698692.1 | - | 1, 2 | 135 | ||