siRNA Details
| Gene Symbol: LOC101926934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134642.1 | 2 | 271 | |||
| NR_134643.1 | 2 | 269 | ||||
| NR_134644.1 | 2 | 196 | ||||
| GenBank mRNA | AA725776.1 | - | 1 | 98 | ||
| AI638322.1 | - | 1 | 113 | |||
| AI698692.1 | - | 1 | 99 | |||
| BX114358.1 | - | 2 | 272 | |||

LOC101926934
Genomic Map