siRNA Details
| Gene Symbol: LOC100131289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038929.1 | 2, 3 | 153 | |||
| GenBank mRNA | BX090544.1 | - | 2, 3 | 159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100131289| Gene Symbol: LOC100131289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038929.1 | 2, 3 | 153 | |||
| GenBank mRNA | BX090544.1 | - | 2, 3 | 159 | ||