siRNA Details
| Gene Symbol: TEX41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033870.2 | 2 | 221 | |||
| GenBank mRNA | BC043549.1 | - | 2 | 162 | ||
| BQ438527.1 | - | 4 | 457 | |||
| BX648102.1 | - | 2 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX41| Gene Symbol: TEX41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033870.2 | 2 | 221 | |||
| GenBank mRNA | BC043549.1 | - | 2 | 162 | ||
| BQ438527.1 | - | 4 | 457 | |||
| BX648102.1 | - | 2 | 204 | |||