siRNA Details
Gene Symbol: TEX41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033870.2 | 2 | 303 | |||
GenBank mRNA | BC043549.1 | - | 2 | 244 | ||
BQ438527.1 | - | 4 | 539 | |||
BX648102.1 | - | 2 | 286 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEX41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033870.2 | 2 | 303 | |||
GenBank mRNA | BC043549.1 | - | 2 | 244 | ||
BQ438527.1 | - | 4 | 539 | |||
BX648102.1 | - | 2 | 286 |