siRNA Details
| Gene Symbol: SHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016848.5 | NP_058544.3 | 12 | 7841 | ||
| XM_011518785.2 | XP_011517087.1 | 12 | 11383 | |||
| XM_011518786.2 | XP_011517088.1 | 13 | 7232 | |||
| GenBank mRNA | AK127258.1 | - | 1 | 1597 | ||
| BC042431.1 | - | 1 | 1944 | |||
| BX538228.1 | - | 1 | 2703 | |||
| Gene Symbol: LOC100131540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129921.1 | - | 1 | 3067 | ||

SHC3
Genomic Map