siRNA Details
| Gene Symbol: LINC00887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024480.1 | 2 | 230 | |||
| GenBank mRNA | AK054628.1 | - | 2 | 233 | ||
| BC112168.1 | - | 2 | 64 | |||
| BC113537.1 | - | 2 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00887| Gene Symbol: LINC00887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024480.1 | 2 | 230 | |||
| GenBank mRNA | AK054628.1 | - | 2 | 233 | ||
| BC112168.1 | - | 2 | 64 | |||
| BC113537.1 | - | 2 | 64 | |||