siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 13 | 2524 | |||
| GenBank mRNA | AL833650.1 | - | 5 | 572 | ||
| BX537595.1 | - | 11 | 2602 | |||
| BX648193.1 | - | 13 | 2520 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 13 | 2524 | |||
| GenBank mRNA | AL833650.1 | - | 5 | 572 | ||
| BX537595.1 | - | 11 | 2602 | |||
| BX648193.1 | - | 13 | 2520 | |||