siRNA Details
| Gene Symbol: FLJ40039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097358.1 | - | 2 | 1762 | ||
| AK127737.1 | - | 2 | 1907 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ40039| Gene Symbol: FLJ40039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097358.1 | - | 2 | 1762 | ||
| AK127737.1 | - | 2 | 1907 | |||