siRNA Details
| Gene Symbol: LOC105377590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133944.1 | 2 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377590| Gene Symbol: LOC105377590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133944.1 | 2 | 384 | |||