siRNA Details
| Gene Symbol: LOC100132071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128253.1 | - | 1 | 845 | ||
| Gene Symbol: CLEC16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011522435.1 | XP_011520737.1 | 24 | 3137 | ||
| GenBank mRNA | AK126771.1 | - | 3 | 351 | ||

LOC100132071
Genomic Map