siRNA Details
| Gene Symbol: LOC101060498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121640.1 | 2 | 644 | |||
| NR_121641.1 | 3 | 500 | ||||
| GenBank mRNA | BM052690.1 | - | 2 | 322 | ||
| JN120857.1 | - | 2 | 392 | |||
| JN120858.1 | - | 3 | 501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101060498| Gene Symbol: LOC101060498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121640.1 | 2 | 644 | |||
| NR_121641.1 | 3 | 500 | ||||
| GenBank mRNA | BM052690.1 | - | 2 | 322 | ||
| JN120857.1 | - | 2 | 392 | |||
| JN120858.1 | - | 3 | 501 | |||