siRNA Details
| Gene Symbol: LOC441666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024380.1 | 1 | 88 | |||
| GenBank mRNA | AF519622.1 | - | 1 | 40 | ||
| AK131313.1 | - | 1 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441666| Gene Symbol: LOC441666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024380.1 | 1 | 88 | |||
| GenBank mRNA | AF519622.1 | - | 1 | 40 | ||
| AK131313.1 | - | 1 | 88 | |||