siRNA Details
| Gene Symbol: RPPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002312.1 | 1 | 270 | |||
| GenBank mRNA | CA413366.1 | - | 1 | 91 | ||
| CV309337.1 | - | 1 | 188 | |||
| X15624.1 | - | 1 | 269 | |||
| Gene Symbol: PARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020912.1 | XP_016876401.1 | 1 | 98 | ||

RPPH1
Genomic Map