siRNA Details
| Gene Symbol: CA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121630.1 | 3 | 427 | |||
| NR_121631.1 | 3 | 199 | ||||
| NR_121632.1 | 3 | 216 | ||||
| GenBank mRNA | BM713112.1 | - | 2 | 107 | ||
| BM717037.1 | - | 2 | 107 | |||
| DA940436.1 | - | 3 | 199 | |||
| DB177224.1 | - | 3 | 286 | |||
| DB196812.1 | - | 3 | 427 | |||

CA3-AS1
Genomic Map