siRNA Details
| Gene Symbol: KDM4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033827.1 | 3, 4 | 468 | |||
| GenBank mRNA | BC094867.1 | - | 3, 4 | 468 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KDM4A-AS1| Gene Symbol: KDM4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033827.1 | 3, 4 | 468 | |||
| GenBank mRNA | BC094867.1 | - | 3, 4 | 468 | ||