siRNA Details
| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC022205.1 | - | 1 | 818 | ||
| KJ782215.1 | - | 2 | 818 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG12| Gene Symbol: SNHG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC022205.1 | - | 1 | 818 | ||
| KJ782215.1 | - | 2 | 818 | |||