siRNA Details
| Gene Symbol: FAM223B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027402.1 | 1 | 487 | |||
| GenBank mRNA | AY168775.1 | - | 1 | 487 | ||
| Gene Symbol: FAM223A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027401.2 | 1 | 487 | |||

FAM223B
Genomic Map