siRNA Details
| Gene Symbol: LOC101928953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094577.1 | - | 2 | 114 | ||
| AK126743.1 | - | 1 | 2152 | |||
| DR761867.1 | - | 2 | 587 | |||
| Gene Symbol: PPP1R16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090471.1 | - | 4 | 1799 | ||

LOC101928953
Genomic Map