siRNA Details
| Gene Symbol: LINC01558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026773.1 | 3 | 423 | |||
| GenBank mRNA | AB016901.1 | - | 3 | 435 | ||
| BC027853.1 | - | 3 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01558| Gene Symbol: LINC01558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026773.1 | 3 | 423 | |||
| GenBank mRNA | AB016901.1 | - | 3 | 435 | ||
| BC027853.1 | - | 3 | 446 | |||