siRNA Details
| Gene Symbol: ASMTL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026710.1 | 4 | 1085 | |||
| GenBank mRNA | BC093994.2 | - | 3 | 428 | ||
| BC143527.1 | - | 3 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASMTL-AS1| Gene Symbol: ASMTL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026710.1 | 4 | 1085 | |||
| GenBank mRNA | BC093994.2 | - | 3 | 428 | ||
| BC143527.1 | - | 3 | 462 | |||