siRNA Details
| Gene Symbol: NBPF25P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104217.1 | 2 | 175 | |||
| GenBank mRNA | AK125792.1 | - | 2 | 175 | ||
| DB049385.1 | - | 2 | 440 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NBPF25P| Gene Symbol: NBPF25P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104217.1 | 2 | 175 | |||
| GenBank mRNA | AK125792.1 | - | 2 | 175 | ||
| DB049385.1 | - | 2 | 440 | |||