siRNA Details
| Gene Symbol: KCNIP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045118.1 | 1 | 112 | |||
| GenBank mRNA | BX090826.1 | - | 1 | 50 | ||
| DB457968.1 | - | 1 | 112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNIP2-AS1| Gene Symbol: KCNIP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045118.1 | 1 | 112 | |||
| GenBank mRNA | BX090826.1 | - | 1 | 50 | ||
| DB457968.1 | - | 1 | 112 | |||