siRNA Details
Gene Symbol: KCNIP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045118.1 | 2 | 198 | |||
GenBank mRNA | BX090826.1 | - | 2 | 136 | ||
DB457968.1 | - | 2 | 198 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNIP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045118.1 | 2 | 198 | |||
GenBank mRNA | BX090826.1 | - | 2 | 136 | ||
DB457968.1 | - | 2 | 198 |