siRNA Details
| Gene Symbol: LOC101927762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120629.1 | 3 | 223 | |||
| GenBank mRNA | AI989559.1 | - | 1 | 178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927762| Gene Symbol: LOC101927762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120629.1 | 3 | 223 | |||
| GenBank mRNA | AI989559.1 | - | 1 | 178 | ||