siRNA Details
Gene Symbol: LOC200772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033841.1 | 6 | 1183 | |||
GenBank mRNA | BC040629.1 | - | 6 | 1183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC200772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033841.1 | 6 | 1183 | |||
GenBank mRNA | BC040629.1 | - | 6 | 1183 |