siRNA Details
| Gene Symbol: SHANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012309.4 | NP_036441.2 | 7 | 934 | ||
| XM_017017387.1 | XP_016872876.1 | 8 | 1039 | |||
| XM_017017388.1 | XP_016872877.1 | 8 | 1061 | |||
| XM_017017389.1 | XP_016872878.1 | 8 | 1234 | |||
| GenBank mRNA | AB208025.1 | - | 7 | 900 | ||
| AK096045.1 | - | 10 | 1111 | |||
| DQ152234.1 | - | 6 | 822 | |||

SHANK2
Genomic Map