siRNA Details
| Gene Symbol: HDAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015053.1 | NP_001015053.1 | 11 | 1590 | ||
| NM_005474.4 | NP_005465.2 | 11 | 1587 | |||
| XM_005256905.1 | XP_005256962.1 | 11 | 1585 | |||
| XM_005256906.4 | XP_005256963.1 | 11 | 1546 | |||
| XM_011524149.1 | XP_011522451.1 | 11 | 1561 | |||
| XM_011524150.1 | XP_011522452.1 | 10 | 1337 | |||
| XM_017023988.1 | XP_016879477.1 | 13 | 1699 | |||
| XM_017023989.1 | XP_016879478.1 | 13 | 1665 | |||
| XM_017023990.1 | XP_016879479.1 | 13 | 1690 | |||
| XM_017023991.1 | XP_016879480.1 | 13 | 1696 | |||
| XM_017023992.1 | XP_016879481.1 | 13 | 1685 | |||
| XM_017023993.1 | XP_016879482.1 | 13 | 1672 | |||
| XM_017023994.1 | XP_016879483.1 | 9 | 1653 | |||
| XM_017023995.1 | XP_016879484.1 | 11 | 1555 | |||
| XM_017023996.1 | XP_016879485.1 | 12 | 1762 | |||
| GenBank mRNA | AB011172.2 | - | 11 | 1559 | ||
| AB177853.1 | - | 1 | 39 | |||
| AF039691.1 | - | 6 | 692 | |||
| AF132608.1 | - | 10 | 1255 | |||
| AK075095.1 | - | 2 | 236 | |||
| AK294760.1 | - | 11 | 1555 | |||
| BC013140.1 | - | 8 | 1037 | |||
| BC051824.1 | - | 11 | 1466 | 6043491 | ||

HDAC5
Genomic Map