siRNA Details
| Gene Symbol: TRIM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006778.3 | NP_006769.2 | 7 | 1074 | ||
| NM_052828.2 | NP_439893.2 | 7 | 1074 | |||
| XM_011514221.1 | XP_011512523.1 | 9 | 1322 | |||
| XM_011514222.2 | XP_011512524.1 | 9 | 1556 | |||
| XM_011514223.2 | XP_011512525.1 | 10 | 1437 | |||
| XM_011514225.1 | XP_011512527.1 | 9 | 1322 | |||
| GenBank mRNA | AF220123.1 | - | 7 | 1072 | ||
| AL360163.1 | - | 5 | 337 | |||
| AL360164.1 | - | 5 | 368 | |||
| BC093926.1 | - | 7 | 1052 | 7939771 | ||
| BC113419.1 | - | 7 | 1052 | 8322471 | ||

TRIM10
Genomic Map