siRNA Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190977.2 | NP_001177906.1 | 2 | 242 | ||
| NM_001190979.2 | NP_001177908.1 | 4 | 440 | |||
| NM_001320080.1 | NP_001307009.1 | 4 | 468 | |||
| NM_005748.5 | NP_005739.2 | 3 | 368 | |||
| NR_135139.1 | 6 | 610 | ||||
| NR_135140.1 | 6 | 642 | ||||
| XM_006719184.3 | XP_006719247.1 | 3 | 287 | |||
| XM_006719185.3 | XP_006719248.1 | 3 | 312 | |||
| XM_011537728.2 | XP_011536030.1 | 5 | 467 | |||
| XM_011537729.2 | XP_011536031.1 | 3 | 257 | |||
| XM_017018669.1 | XP_016874158.1 | 5 | 426 | |||
| GenBank mRNA | AI654632.1 | - | 1 | 64 | ||
| AK291355.1 | - | 3 | 248 | |||
| AK294260.1 | - | 2 | 163 | |||
| AK296191.1 | - | 4 | 348 | |||
| BC037777.2 | - | 4 | 308 | 5276663 | ||
| CB963485.1 | - | 6 | 497 | |||
| CB993159.1 | - | 6 | 489 | |||
| DA011078.1 | - | 3 | 349 | |||
| DA491895.1 | - | 3 | 352 | |||

YAF2
Genomic Map