siRNA Details
| Gene Symbol: ANGPTL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021146.3 | NP_066969.1 | 1, 2 | 665 | ||
| XM_017000004.1 | XP_016855493.1 | 1, 2 | 630 | |||
| GenBank mRNA | AB593026.1 | - | 1, 2 | 599 | ||
| AB593027.1 | - | 1, 2 | 666 | |||
| AK313716.1 | - | 1, 2 | 631 | |||
| AY358301.1 | - | 1, 2 | 588 | |||
| BC001881.1 | - | 1, 2 | 613 | 3544149 | ||
| BT009802.1 | - | 1, 2 | 374 | |||
| Y16132.1 | - | 1, 2 | 381 | |||

ANGPTL7
Genomic Map