siRNA Details
| Gene Symbol: MFSD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001120.4 | NP_001111.3 | 3 | 605 | ||
| NM_001146069.1 | NP_001139541.1 | 4 | 403 | |||
| XM_005247927.3 | XP_005247984.1 | 4 | 398 | |||
| GenBank mRNA | BC001502.2 | - | 4 | 329 | 2988084 | |
| BC014979.1 | - | 4 | 332 | 4878602 | ||
| BI769550.1 | - | 3 | 602 | |||
| BX359190.2 | - | 4 | 403 | |||
| L11669.1 | - | 3 | 411 | |||

MFSD10
Genomic Map