siRNA Details
| Gene Symbol: LRRC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135217.1 | NP_001128689.1 | 5 | 925 | ||
| NM_006992.3 | NP_008923.1 | 5 | 753 | |||
| NM_201650.2 | NP_964013.1 | 5 | 753 | |||
| GenBank mRNA | AI524335.1 | - | 2 | 459 | ||
| AK223535.1 | - | 5 | 687 | |||
| AK292291.1 | - | 5 | 688 | |||
| AK292367.1 | - | 5 | 753 | |||
| BC029858.1 | - | 5 | 726 | 5170643 | ||
| CA444672.1 | - | 2 | 443 | |||
| U72508.1 | - | 5 | 645 | |||

LRRC23
Genomic Map