siRNA Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005858.3 | NP_005849.1 | 13 | 1634 | ||
| XM_011527624.2 | XP_011525926.1 | 13 | 1727 | |||
| XM_017026141.1 | XP_016881630.1 | 13 | 1779 | |||
| GenBank mRNA | AK309529.1 | - | 10 | 1162 | ||
| AL050160.1 | - | 7 | 562 | |||
| BC037270.2 | - | 13 | 1624 | 4837309 | ||
| Y11997.1 | - | 13 | 1625 | |||

AKAP8
Genomic Map