siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 6, 7 | 668 | ||
| NM_006061.2 | NP_006052.2 | 6, 7 | 656 | |||
| GenBank mRNA | AK223380.1 | - | 6, 7 | 560 | ||
| AK292786.1 | - | 6, 7 | 615 | |||
| AK303966.1 | - | 4, 5 | 383 | |||
| AK313387.1 | - | 6, 7 | 589 | |||
| BC069580.1 | - | 5, 6 | 542 | 7262396 | ||
| BC069602.1 | - | 5, 6 | 544 | 7262408 | ||
| BC101539.1 | - | 5, 6 | 566 | 8069045 | ||
| X94323.1 | - | 6, 7 | 569 | |||
| X95240.1 | - | 5, 6 | 544 | |||

CRISP3
Genomic Map