siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 776 | ||
| NM_006061.2 | NP_006052.2 | 8 | 764 | |||
| GenBank mRNA | AK223380.1 | - | 8 | 668 | ||
| AK292786.1 | - | 8 | 723 | |||
| AK303966.1 | - | 6 | 491 | |||
| AK313387.1 | - | 8 | 697 | |||
| BC069580.1 | - | 7 | 650 | 7262396 | ||
| BC069602.1 | - | 7 | 652 | 7262408 | ||
| BC101539.1 | - | 7 | 674 | 8069045 | ||
| X94323.1 | - | 8 | 677 | |||
| X95240.1 | - | 7 | 652 | |||

CRISP3
Genomic Map