siRNA Details
| Gene Symbol: KLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016270.3 | NP_057354.1 | 2 | 953 | ||
| GenBank mRNA | AF134053.1 | - | 2 | 855 | ||
| AF205849.1 | - | 2 | 939 | |||
| AK225820.1 | - | 2 | 952 | |||
| BC071983.1 | - | 2 | 942 | 5480246 | ||

KLF2
Genomic Map