siRNA Details
| Gene Symbol: TUBB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289123.1 | NP_001276052.1 | 3 | 427 | ||
| NM_001289127.1 | NP_001276056.1 | 3 | 409 | |||
| NM_001289129.1 | NP_001276058.1 | 3 | 357 | |||
| NM_001289130.1 | NP_001276059.1 | 2 | 466 | |||
| NM_001289131.1 | NP_001276060.1 | 2 | 433 | |||
| NM_006087.3 | NP_006078.2 | 2 | 509 | |||
| GenBank mRNA | AK075307.1 | - | 2 | 235 | ||
| AK095202.1 | - | 1 | 766 | |||
| AK293503.1 | - | 2 | 158 | |||
| AK295917.1 | - | 2 | 234 | |||
| BC006570.2 | - | 2 | 218 | 3603726 | ||
| BC013683.1 | - | 2 | 244 | 4155617 | ||
| DA265791.1 | - | 2 | 194 | |||
| DA302559.1 | - | 2 | 187 | |||
| DC303150.1 | - | 3 | 382 | |||
| DC314277.1 | - | 2 | 509 | |||
| DC353527.1 | - | 3 | 357 | |||
| DC354036.1 | - | 3 | 360 | |||

TUBB4A
Genomic Map