siRNA Details
| Gene Symbol: CDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001802.1 | NP_001793.1 | 5 | 1214 | ||
| GenBank mRNA | AK292273.1 | - | 5 | 961 | ||
| BC017503.1 | - | 5 | 1196 | 4873337 | ||
| D12981.1 | - | 5 | 890 | |||
| M63256.1 | - | 5 | 1074 | |||

CDR2
Genomic Map