siRNA Details
| Gene Symbol: WFDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006103.3 | NP_006094.3 | 1 | 86 | ||
| GenBank mRNA | AF330259.1 | - | 1 | 75 | ||
| AF330260.1 | - | 1 | 75 | |||
| AF330262.1 | - | 1 | 75 | |||
| AK225528.1 | - | 1 | 85 | |||
| AK290288.1 | - | 1 | 87 | |||
| AK311966.1 | - | 1 | 87 | |||
| AY212888.1 | - | 1 | 95 | |||
| BC039816.1 | - | 1 | 63 | |||
| BC046106.1 | - | 1 | 62 | 5186932 | ||
| BI759775.1 | - | 1 | 114 | |||
| CR456977.1 | - | 1 | 58 | |||
| X63187.1 | - | 1 | 85 | |||

WFDC2
Genomic Map