siRNA Details
| Gene Symbol: COG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161520.1 | NP_001154992.1 | 5 | 1018 | ||
| NM_006348.3 | NP_006339.3 | 5 | 1018 | |||
| NM_181733.2 | NP_859422.2 | 5 | 1018 | |||
| GenBank mRNA | AF058718.1 | - | 5 | 544 | ||
| AK314270.1 | - | 5 | 1019 | |||
| BC068540.1 | - | 5 | 765 | 4374289 | ||
| BI597200.1 | - | 5 | 428 | |||

COG5
Genomic Map