siRNA Details
| Gene Symbol: SSSCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303024.1 | NP_001289953.1 | 3 | 553 | ||
| NM_006396.2 | NP_006387.1 | 4 | 460 | |||
| GenBank mRNA | AB001740.1 | - | 4 | 447 | ||
| BC014791.2 | - | 4 | 446 | 3857068 | ||
| BP351453.1 | - | 4 | 461 | |||
| DR759640.1 | - | 3 | 520 | |||

SSSCA1
Genomic Map