siRNA Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006426.2 | NP_006417.2 | 1, 2 | 196 | ||
| XM_011539125.2 | XP_011537427.1 | 1, 2 | 47 | |||
| GenBank mRNA | AB006713.1 | - | 1, 2 | 190 | ||
| BC029280.1 | - | 1, 2 | 80 | |||
| BC136329.1 | - | 1, 2 | 65 | 9020316 | ||
| BC136330.1 | - | 1, 2 | 65 | 9020317 | ||
| Y10976.1 | - | 1, 2 | 73 | |||

DPYSL4
Genomic Map