siRNA Details
| Gene Symbol: PDPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006624.1 | NP_001006625.1 | 3 | 345 | ||
| NM_001006625.1 | NP_001006626.1 | 3 | 345 | |||
| NM_006474.4 | NP_006465.3 | 3 | 521 | |||
| NM_198389.2 | NP_938203.2 | 3 | 521 | |||
| XM_006710295.1 | XP_006710358.1 | 3 | 252 | |||
| GenBank mRNA | AB127958.1 | - | 3 | 244 | ||
| AF030428.1 | - | 3 | 424 | |||
| AF390106.1 | - | 3 | 427 | |||
| AJ225022.1 | - | 3 | 376 | |||
| AK075327.1 | - | 3 | 446 | |||
| AK075345.1 | - | 3 | 446 | |||
| AK307705.1 | - | 3 | 252 | |||
| AK307706.1 | - | 4 | 407 | |||
| AK312607.1 | - | 3 | 447 | |||
| AY194238.1 | - | 3 | 447 | |||
| BC014668.1 | - | 3 | 477 | 4876446 | ||
| BC022812.1 | - | 3 | 477 | 4875444 | ||
| BE887331.1 | - | 523 | ||||
| BQ212176.1 | - | 3 | 342 | |||
| BQ214601.1 | - | 3 | 274 | |||
| CA312590.1 | - | 4 | 593 | |||

PDPN
Genomic Map