siRNA Details
| Gene Symbol: EXOC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006544.3 | NP_006535.1 | 11 | 1215 | ||
| XM_005267272.3 | XP_005267329.1 | 11 | 1300 | |||
| GenBank mRNA | AK295085.1 | - | 11 | 1122 | ||
| AK303531.1 | - | 10 | 958 | |||
| AK312523.1 | - | 11 | 1220 | |||
| AL833745.1 | - | 1 | 682 | |||
| BC041126.1 | - | 11 | 1181 | 3914527 | ||
| U85946.1 | - | 11 | 1159 | |||

EXOC5
Genomic Map