siRNA Details
| Gene Symbol: CGRRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006568.2 | NP_006559.1 | 1, 2 | 185 | ||
| GenBank mRNA | AK289478.1 | - | 1, 2 | 216 | ||
| AK315318.1 | - | 1, 2 | 226 | |||
| BC015063.1 | - | 1, 2 | 151 | 4245551 | ||
| BG701924.1 | - | 1, 2 | 246 | |||
| U66469.1 | - | 1, 2 | 121 | |||

CGRRF1
Genomic Map